accessibility menu, dialog, popup

UserName

CAS RN 26524-88-9

IUPAC Name: 2-(benzylsulfanyl)ethan-1-ol
Synonyms: 2-(benzylsulfanyl)ethanol
Molecular Weight (g/mol): 168.25
Molecular Formula: C9H12OS
InChi Key: ANMHSIXPUAKNLM-UHFFFAOYSA-N
SMILES: OCCSCC1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)