missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxyethanol, 98%

Catalog No. AC314650250
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314650250 25 g Glass bottle
AC314650010 1 g Glass bottle
AC314650050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC314650250 Supplier Thermo Scientific Chemicals Supplier No. 314650250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 622-08-2 | C9H12O2 | 152.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 622-08-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002868
InChI Key CUZKCNWZBXLAJX-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
PubChem CID 12141
IUPAC Name 2-phenylmethoxyethanol
SMILES OCCOCC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Boiling Point 265°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.522
Linear Formula C6H5CH2OCH2CH2OH
Quantity 25 g
Specific Gravity 1.07
Formula Weight 152.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Benzyloxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-719-3

RUO – Research Use Only