accessibility menu, dialog, popup

UserName

HOO

CAS RN 622-08-2

IUPAC Name: 2-(benzyloxy)ethan-1-ol
Synonyms: benzyl glycol
Molecular Weight (g/mol): 152.19
Molecular Formula: C9H12O2
InChi Key: CUZKCNWZBXLAJX-UHFFFAOYSA-N
SMILES: OCCOCC1=CC=CC=C1
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)