Learn More
2-Benzyloxybenzoic acid, 98%
CAS: 14389-86-7 | C14H11O3 | 227.24 g/mol
Supplier: Thermo Scientific Chemicals A1242518
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Benzyloxybenzoic acid | |
| 76°C to 78°C | |
| MFCD00051940 | |
| 2-benzyloxy benzoic acid, 2-benzyloxybenzoic acid, benzoic acid,2-phenylmethoxy, 2-phenylmethoxy benzoic acid, benzoic acid, 2-phenylmethoxy, enamine_005339, benzyloxybenzoic acid, acmc-20ao3m, 2-benzyloxy-benzoic acid, 2-benzyloxybenzoicacid | |
| [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1 | |
| 227.24 | |
| 228.25 |
| 14389-86-7 | |
| C14H11O3 | |
| 50 g | |
| GMOYUTKNPLBTMT-UHFFFAOYSA-M | |
| 2-(benzyloxy)benzoate | |
| 1810581 | |
| 98% |
Chemical Identifiers
| 14389-86-7 | |
| 227.24 | |
| GMOYUTKNPLBTMT-UHFFFAOYSA-M | |
| [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1 |
| C14H11O3 | |
| MFCD00051940 | |
| 1810581 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only