missing translation for 'onlineSavingsMsg'
Learn More

2-Benzyloxybenzoic acid, 98%

Catalog No. p-7082137
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1242509 10 g
AAA1242518 50 g
AAA1242530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1242509 Supplier Thermo Scientific Chemicals Supplier No. A1242509
Only null left

CAS: 14389-86-7 | C14H11O3 | 227.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14389-86-7
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00051940
InChI Key GMOYUTKNPLBTMT-UHFFFAOYSA-M
PubChem CID 1810581
SMILES [O-]C(=O)C1=CC=CC=C1OCC1=CC=CC=C1

Specifications

Melting Point 76°C to 78°C
Quantity 10 g
Synonym 2-benzyloxy benzoic acid, 2-benzyloxybenzoic acid, benzoic acid,2-phenylmethoxy, 2-phenylmethoxy benzoic acid, benzoic acid, 2-phenylmethoxy, enamine_005339, benzyloxybenzoic acid, acmc-20ao3m, 2-benzyloxy-benzoic acid, 2-benzyloxybenzoicacid
IUPAC Name 2-phenylmethoxybenzoic acid
Formula Weight 228.25
Percent Purity 98%
Chemical Name or Material 2-Benzyloxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only