missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)phenol 97.0+%, TCI America™

Catalog No. B166110G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B166110G 10 g
B166125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B166110G Supplier TCI America Supplier No. B166110G

Chemical Identifiers

CAS 6272-38-4
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00002186
InChI Key CCZCXFHJMKINPE-UHFFFAOYSA-N
Synonym 2-benzyloxy phenol, 2-benzyloxyphenol, phenol, 2-phenylmethoxy, benzyloxyphenol, benzyl o-hydroxyphenyl ether, 2-phenylmethoxy phenol, 2-benzyloxy-phenol, o-benzyloxy phenol
PubChem CID 80459
IUPAC Name 2-phenylmethoxyphenol
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2O

Specifications

Color Yellow
Boiling Point 215°C
Quantity 10 g
Formula Weight 200.24
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Benzyloxy)phenol