missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)ethanol 98.0+%, TCI America™

Catalog No. p-7118440
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B042625ML 25 mL
B0426250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B042625ML Supplier TCI America Supplier No. B042625ML

Chemical Identifiers

CAS 622-08-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002868
InChI Key CUZKCNWZBXLAJX-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
PubChem CID 12141
IUPAC Name 2-(benzyloxy)ethan-1-ol
SMILES OCCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 256°C
Quantity 25 mL
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Benzyloxy)ethanol
RTECSNumber KJ7550000
TSCA Yes