missing translation for 'onlineSavingsMsg'
Learn More

2-(Benzyloxy)ethanol 98.0+%, TCI America™

Catalog No. b0426250ml
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
250 mL
Catalog No. Quantity
B0426250ML 250 mL
B042625ML 25 mL
2 options

Catalog No. B0426250ML

Supplier: TCI America B0426250ML

Only null left

Chemical Identifiers

CAS 622-08-2
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00002868
InChI Key CUZKCNWZBXLAJX-UHFFFAOYSA-N
Synonym 2-benzyloxy ethanol, 2-benzyloxyethanol, ethanol, 2-phenylmethoxy, ethylene glycol monobenzyl ether, 2-benzyloxy-1-ethanol, benzyl cellosolve, glycol benzyl ether, ethanol, 2-benzyloxy, glycol monobenzyl ether
PubChem CID 12141
IUPAC Name 2-(benzyloxy)ethan-1-ol
SMILES OCCOCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 256°C
Quantity 250 mL
Formula Weight 152.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(Benzyloxy)ethanol
RTECSNumber KJ7550000
TSCA Yes