missing translation for 'onlineSavingsMsg'
Learn More

2-Anthraceneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A18431G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A18431G Supplier TCI America Supplier No. A18431G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 141981-64-8
Molecular Formula C14H11BO2
Molecular Weight (g/mol) 222.05
MDL Number MFCD07784001
InChI Key PKWBMOXZIMVOJT-UHFFFAOYSA-N
PubChem CID 9855952
IUPAC Name (anthracen-2-yl)boronic acid
SMILES OB(O)C1=CC=C2C=C3C=CC=CC3=CC2=C1

Specifications

Quantity 1 g
Formula Weight 222.05
Physical Form Crystalline Powder
Chemical Name or Material 2-Anthraceneboronic Acid (contains varying amounts of Anhydride)