accessibility menu, dialog, popup

UserName

CAS RN 141981-64-8

IUPAC Name: (anthracen-2-yl)boronic acid
Synonyms: anthracen-2-ylboronic acid
Molecular Weight (g/mol): 222.05
Molecular Formula: C14H11BO2
InChi Key: PKWBMOXZIMVOJT-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=C3C=CC=CC3=CC2=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results