missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiobenzamide, 97%

Catalog No. AAH5180903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5180903 1 g
AAH5180906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5180903 Supplier Thermo Scientific Chemicals Supplier No. H5180903

CAS: 2454-39-9 | C7H8N2S | 152.215 g/mol

It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be heated to produce 2-trifluoromethyl-3H-quinazoline-4-thione in the presence of reagent molecular sieves and solvent CHCl3 with reaction time of 2 hours. It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 2454-39-9
Molecular Formula C7H8N2S
Molecular Weight (g/mol) 152.215
MDL Number MFCD00963496
InChI Key HPZKAJRFABCGFF-UHFFFAOYSA-N
Synonym 2-aminothiobenzamide, 2-amino-thiobenzamide, 2-aminobenzothioamide, benzenecarbothioamide, 2-amino, thioanthranilamide, o-aminothiobenzamide, 2-aminobenzene-1-carbothioamide, amino 2-aminophenyl methane-1-thione, benzamide, o-aminothio, thioanthranilsaureamid
PubChem CID 1550990
IUPAC Name 2-aminobenzenecarbothioamide
SMILES C1=CC=C(C(=C1)C(=S)N)N

Specifications

Melting Point 115°C to 120°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 152.21
Percent Purity 97%
Chemical Name or Material 2-Aminothiobenzamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only