Learn More
2-Aminothiazole-5-carboxylic acid, 95%, Thermo Scientific™
$362.12 - $1,062.99
Chemical Identifiers
| CAS | 40283-46-3 |
|---|---|
| Molecular Formula | C4H4N2O2S |
| Molecular Weight (g/mol) | 144.15 |
| MDL Number | MFCD06203554 |
| InChI Key | ZFMRDDYYJJCBKC-UHFFFAOYSA-N |
| Synonym | 2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid |
| PubChem CID | 315243 |
| IUPAC Name | 2-amino-1,3-thiazole-5-carboxylic acid |
| SMILES | C1=C(SC(=N1)N)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC429570010
|
N/A | 1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC429570050
|
N/A | 5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 40283-46-3 | |
| 144.15 | |
| ZFMRDDYYJJCBKC-UHFFFAOYSA-N | |
| 315243 | |
| C1=C(SC(=N1)N)C(=O)O |
| C4H4N2O2S | |
| MFCD06203554 | |
| 2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid | |
| 2-amino-1,3-thiazole-5-carboxylic acid |
Specifications
| 40283-46-3 | |
| 100.0 | |
| 94% min. (HPLC) | |
| MFCD06203554 | |
| ZFMRDDYYJJCBKC-UHFFFAOYSA-N | |
| 2-amino-1,3-thiazole-5-carboxylic acid | |
| 315243 | |
| 95% |
| 94.0 | |
| Authentic | |
| C4H4N2O2S | |
| 2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid | |
| C1=C(SC(=N1)N)C(=O)O | |
| 144.15 | |
| 144.15 | |
| 2-Aminothiazole-5-carboxylic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash
GHS Signal Word: Warning
RUO – Research Use Only