missing translation for 'onlineSavingsMsg'
Learn More

2-Aminothiazole-5-carboxylic acid, 95%

Catalog No. AC429570010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429570010 1 g Glass bottle
AC429570050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429570010 Supplier Thermo Scientific Chemicals Supplier No. 429570010

CAS: 40283-46-3 | C4H4N2O2S | 144.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40283-46-3
Molecular Formula C4H4N2O2S
Molecular Weight (g/mol) 144.15
MDL Number MFCD06203554
InChI Key ZFMRDDYYJJCBKC-UHFFFAOYSA-N
Synonym 2-aminothiazole-5-carboxylic acid, 2-amino-5-thiazolecarboxylic acid, 5-thiazolecarboxylic acid, 2-amino, 2-amino-5-carboxy-1,3-thiazole, pubchem21384, acmc-209jcp, 2-aminothiazole-5-carboxylicacid, 5-thiazolecarboxylicacid, 2-amino, 2-aminothiazole-5-formic acid, 2-azanyl-1,3-thiazole-5-carboxylic acid
PubChem CID 315243
IUPAC Name 2-amino-1,3-thiazole-5-carboxylic acid
SMILES C1=C(SC(=N1)N)C(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 182.0°C to 184.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 144.15
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminothiazole-5-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

RUO – Research Use Only