accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 40283-46-3

NH2SNOHO

CAS RN 40283-46-3

IUPAC Name: 2-amino-1,3-thiazole-5-carboxylic acid
Synonyms: 2-amino-1,3-thiazole-5-carboxylic acid
Molecular Weight (g/mol): 144.15
Molecular Formula: C4H4N2O2S
InChi Key: ZFMRDDYYJJCBKC-UHFFFAOYSA-N
SMILES: NC1=NC=C(S1)C(O)=O

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.