Learn More
2-Aminophenol, 99%
CAS: 95-55-6 | C6H7NO | 109.128 g/mol
Supplier: Thermo Scientific Chemicals A1373536
Description
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.
Solubility
Soluble in water (17 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
| 2-Aminophenol | |
| 173°C to 177°C | |
| 150°C (11 mmHg) | |
| Characteristic | |
| MFCD00007690 | |
| UN2512 | |
| 14,461 | |
| Soluble in water (17g/L at 20°C). | |
| C1=CC=C(C(=C1)N)O | |
| 109.128 | |
| CHEBI:18112 | |
| 99% |
| 95-55-6 | |
| 1.328 | |
| >175°C (347°F) | |
| C6H7NO | |
| 500 g | |
| 606075 | |
| o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga | |
| CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
| 2-aminophenol | |
| 5801 | |
| 109.13 |
Chemical Identifiers
| 95-55-6 | |
| 109.128 | |
| CDAWCLOXVUBKRW-UHFFFAOYSA-N | |
| 5801 | |
| 2-aminophenol |
| C6H7NO | |
| MFCD00007690 | |
| o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga | |
| CHEBI:18112 | |
| C1=CC=C(C(=C1)N)O |
Safety and Handling
GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c
H302+H332-H317-H341
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)
EINECSNumber : 202-431-1
RTECSNumber : SJ4950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only