missing translation for 'onlineSavingsMsg'
Learn More

2-Aminophenol, 99%

Catalog No. AC104260050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104260050 5 g Glass bottle
AC104261000 100 g Glass bottle
AC104265000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC104260050 Supplier Thermo Scientific Chemicals Supplier No. 104260050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95-55-6 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 172°C to 177°C
Color White to Brown
Boiling Point 164°C (15.0 Mbar)
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NC6H4OH
Quantity 5 g
Beilstein 13, 354
Merck Index 15, 457
Solubility Information Solubility in water: 17g/L (20°C). Other solubilities: soluble in ether,1g in 23 mL alcohol,slightly soluble in benzene
Formula Weight 109.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Aminophenol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:
EINECSNumber 202-431-1
RTECSNumber SJ4950000
TSCA TSCA

RUO – Research Use Only