missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminodiphenyl Ether 98.0+%, TCI America™
$61.51 - $553.62
Chemical Identifiers
| CAS | 2688-84-8 |
|---|---|
| Molecular Formula | C12H11NO |
| Molecular Weight (g/mol) | 185.226 |
| MDL Number | MFCD00035765 |
| InChI Key | NMFFUUFPJJOWHK-UHFFFAOYSA-N |
| Synonym | 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline |
| PubChem CID | 75899 |
| IUPAC Name | 2-phenoxyaniline |
| SMILES | C1=CC=C(C=C1)OC2=CC=CC=C2N |
Chemical Identifiers
| 2688-84-8 | |
| 185.226 | |
| NMFFUUFPJJOWHK-UHFFFAOYSA-N | |
| 75899 | |
| C1=CC=C(C=C1)OC2=CC=CC=C2N |
| C12H11NO | |
| MFCD00035765 | |
| 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline | |
| 2-phenoxyaniline |
Specifications
| 2688-84-8 | |
| 187°C | |
| MFCD00035765 | |
| 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline | |
| C1=CC=C(C=C1)OC2=CC=CC=C2N | |
| 185.226 | |
| 185.23 | |
| Crystalline Powder |
| 46°C | |
| C12H11NO | |
| 25 g | |
| NMFFUUFPJJOWHK-UHFFFAOYSA-N | |
| 2-phenoxyaniline | |
| 75899 | |
| ≥98.0% (GC,T) | |
| 2-Aminodiphenyl Ether |
Safety and Handling
RTECSNumber : BY7940000
TSCA : Yes
Recommended Storage : Refrigerator