missing translation for 'onlineSavingsMsg'
Learn More

2-Aminodiphenyl Ether 98.0+%, TCI America™

Catalog No. A114225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A114225G 25 g
A1142500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A114225G Supplier TCI America Supplier No. A114225G

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00035765
InChI Key NMFFUUFPJJOWHK-UHFFFAOYSA-N
Synonym 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
PubChem CID 75899
IUPAC Name 2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2N

Specifications

Melting Point 46°C
Boiling Point 187°C
Quantity 25 g
Formula Weight 185.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminodiphenyl Ether
RTECSNumber BY7940000
TSCA Yes
Recommended Storage Refrigerator