Learn More
2-Aminobenzyl alcohol, 98%
CAS: 5344-90-1 | C7H9NO | 123.155 g/mol
Supplier: Thermo Scientific Chemicals A1318030
Description
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2Â to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2Â to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.
Solubility
Very soluble in water.
Notes
Store at room temperature. Incompatible with oxidizing agents.
Specifications
| 2-Aminobenzyl alcohol | |
| 81°C to 84°C | |
| C7H9NO | |
| 250 g | |
| Air and light sensitive | |
| Very soluble in water. | |
| C1=CC=C(C(=C1)CO)N | |
| 123.155 | |
| 123.16 |
| 5344-90-1 | |
| 160°C (10 mmHg) | |
| MFCD00007749 | |
| 1072211 | |
| 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline | |
| VYFOAVADNIHPTR-UHFFFAOYSA-N | |
| (2-aminophenyl)methanol | |
| 21439 | |
| 98% |
Chemical Identifiers
| 5344-90-1 | |
| 123.155 | |
| VYFOAVADNIHPTR-UHFFFAOYSA-N | |
| 21439 | |
| C1=CC=C(C(=C1)CO)N |
| C7H9NO | |
| MFCD00007749 | |
| 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline | |
| (2-aminophenyl)methanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 226-293-7
RTECSNumber : DN3155000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only