accessibility menu, dialog, popup

UserName

CAS RN 5344-90-1

IUPAC Name: (2-aminophenyl)methanol
Synonyms: isatoic anhydride M-aminobenzyl alcohol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: VYFOAVADNIHPTR-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1CO
Molecular Weight (g/mol) 
  • (3)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (5)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)