missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzyl alcohol, 98%

Catalog No. AAA1318009
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1318009 10 g
AAA1318018 50 g
AAA1318030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1318009 Supplier Thermo Scientific Chemicals Supplier No. A1318009
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5344-90-1 | C7H9NO | 123.155 g/mol

2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzyl alcohol is oxidatively cyclised with an array of ketones in dioxane at 80°C in the presence of a ruthenium catalyst and KOH to give corresponding quinolines. It undergoes oxidation catalyzed by heterotrimetallic RuMnMn species on the hydrotalcite surface in the presence of O2 to yield 2-aminobenzaldehyde. was used in the synthesis of ethyl 2-hydroxymethylcarbanilate.

Solubility
Very soluble in water.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5344-90-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
MDL Number MFCD00007749
InChI Key VYFOAVADNIHPTR-UHFFFAOYSA-N
Synonym 2-aminobenzyl alcohol, 2-aminophenyl methanol, 2-aminobenzylalcohol, o-aminobenzyl alcohol, 2-aminobenzenemethanol, benzenemethanol, 2-amino, o-aminobenzylic alcohol, o-hydroxymethyl aniline, benzyl alcohol, o-amino, 2-hydroxymethylaniline
PubChem CID 21439
IUPAC Name (2-aminophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)N

Specifications

Melting Point 81°C to 84°C
Boiling Point 160°C (10 mmHg)
Quantity 10 g
Beilstein 1072211
Sensitivity Air and light sensitive
Solubility Information Very soluble in water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Aminobenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 226-293-7
RTECSNumber DN3155000
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only