missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzotrifluoride 98.0+%, TCI America™

Catalog No. p-7116898
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
A043325ML 25 mL
A0433500ML 500 mL
2 options

Catalog No. A043325ML

Supplier: TCI America A043325ML

Only null left

Chemical Identifiers

CAS 88-17-5
Molecular Formula C7H6F3N
Molecular Weight (g/mol) 161.127
MDL Number MFCD00007718
InChI Key VBLXCTYLWZJBKA-UHFFFAOYSA-N
Synonym 2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
PubChem CID 6922
IUPAC Name 2-(trifluoromethyl)aniline
SMILES C1=CC=C(C(=C1)C(F)(F)F)N

Specifications

Boiling Point 171°C
Quantity 25 mL
UN Number 2942
Formula Weight 161.13
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Aminobenzotrifluoride
EINECSNumber (3)-0223
TSCA Yes