missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzotrifluoride, 99%

CAS: 88-17-5 | C7H6F3N | 161.13 g/mol

Supplier:  Thermo Scientific Chemicals 103181000

 View more versions of this product

Catalog No. AC103181000


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Aminobenzotrifluoride
88-17-5
100.0
Amber to Colorless
170.0°C to 173.0°C (15.0 mmHg)
Authentic
Glass bottle
1.4800 to 1.4820
MFCD00007718
12, II, 453
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
VBLXCTYLWZJBKA-UHFFFAOYSA-N
2-(trifluoromethyl)aniline
6922
99%
99%
98.5
-33.0°C
1.2960g/mL
65°C
98.5% min. (GC)
C7H6F3N
H2NC6H4CF3
100 mL
1.296
Solubility in water: 4g/L in water (20°C). Other solubilities: soluble in ether,ethanol and diethyl ether
C1=CC=C(C(=C1)C(F)(F)F)N
161.13
161.13
Liquid

Chemical Identifiers

88-17-5
161.13
VBLXCTYLWZJBKA-UHFFFAOYSA-N
6922
C1=CC=C(C(=C1)C(F)(F)F)N
C7H6F3N
MFCD00007718
2-trifluoromethyl aniline, 2-aminobenzotrifluoride, o-aminobenzotrifluoride, o-trifluoromethyl aniline, o-trifluoromethylaniline, benzenamine, 2-trifluoromethyl, 2-trifluoromethyl benzenamine, alpha,alpha,alpha-trifluoro-o-toluidine, 2-aminotrifluorotoluene, 2-aminotrifluoromethylbenzene
2-(trifluoromethyl)aniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic lif

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plen

GHS Signal Word: Warning

EINECSNumber : 201-806-7

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only