missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzothiazole, 98%

CAS: 136-95-8 | C7H6N2S | 150.199 g/mol

$79.96 - $284.22

Chemical Identifiers

CAS 136-95-8
Molecular Formula C7H6N2S
Molecular Weight (g/mol) 150.199
MDL Number MFCD00005785
InChI Key UHGULLIUJBCTEF-UHFFFAOYSA-N
Synonym 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
PubChem CID 8706
IUPAC Name 1,3-benzothiazol-2-amine
SMILES C1=CC=C2C(=C1)N=C(S2)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1352122
View Documents
Thermo Scientific Chemicals
A1352122
100 g
Each for $79.96
Only null left
 
AAA1352136
View Documents
Thermo Scientific Chemicals
A1352136
500 g
Each for $284.22
Only null left
 
Description

Description

2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy

Solubility
Soluble in alcohol, chloroform, diethyl ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

136-95-8
150.199
UHGULLIUJBCTEF-UHFFFAOYSA-N
8706
C1=CC=C2C(=C1)N=C(S2)N
C7H6N2S
MFCD00005785
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
1,3-benzothiazol-2-amine

Specifications

136-95-8
C7H6N2S
100 g
14,424
Soluble in alcohol,chloroform,diethyl ether.
C1=CC=C2C(=C1)N=C(S2)N
150.199
150.2
2-Aminobenzothiazole
126°C to 129°C
MFCD00005785
116315
2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine
UHGULLIUJBCTEF-UHFFFAOYSA-N
1,3-benzothiazol-2-amine
8706
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

EINECSNumber : 205-268-4

RTECSNumber : DL1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only