Learn More
2-Aminobenzothiazole, 98%
CAS: 136-95-8 | C7H6N2S | 150.199 g/mol
Supplier: Thermo Scientific Chemicals A1352136
Description
2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminobenzothiazole was used in the synthesis of cobalt(II) picrate mixed-ligand complexes. It was used to study adsorption of biologically significant 2-aminobenzothiazole molecules on colloidal silver particles using surface-enhanced raman scattering spectroscopy
Solubility
Soluble in alcohol, chloroform, diethyl ether.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| 2-Aminobenzothiazole | |
| 126°C to 129°C | |
| MFCD00005785 | |
| 116315 | |
| 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine | |
| UHGULLIUJBCTEF-UHFFFAOYSA-N | |
| 1,3-benzothiazol-2-amine | |
| 8706 | |
| 98% |
| 136-95-8 | |
| C7H6N2S | |
| 500 g | |
| 14,424 | |
| Soluble in alcohol,chloroform,diethyl ether. | |
| C1=CC=C2C(=C1)N=C(S2)N | |
| 150.199 | |
| 150.2 |
Chemical Identifiers
| 136-95-8 | |
| 150.199 | |
| UHGULLIUJBCTEF-UHFFFAOYSA-N | |
| 8706 | |
| C1=CC=C2C(=C1)N=C(S2)N |
| C7H6N2S | |
| MFCD00005785 | |
| 2-aminobenzothiazole, 2-benzothiazolamine, benzo d thiazol-2-amine, benzothiazol-2-ylamine, 2-aminobenzthiazole, benzothiazole-2-ylamine, 2-iminobenzothiazoline, o-aminobenzothiazole, benzothiazole, 2-amino, 2-benzothiazolylamine | |
| 1,3-benzothiazol-2-amine |
Safety and Handling
GHS H Statement
H301-H319
Toxic if swallowed.
Causes serious eye irritation.
GHS P Statement
P280-P301+P310-P305+P351+P338-P321-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
EINECSNumber : 205-268-4
RTECSNumber : DL1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only