accessibility menu, dialog, popup

UserName

H2NSN

CAS RN 136-95-8

IUPAC Name: 1,3-benzothiazol-2-amine
Synonyms: 2-aminobenzothiazole
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: UHGULLIUJBCTEF-UHFFFAOYSA-N
SMILES: NC1=NC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Physical Form 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)