missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminobenzophenone-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. A19941G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19941G 1 g
A19945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19941G Supplier TCI America Supplier No. A19941G

Chemical Identifiers

CAS 1147-43-9
Molecular Formula C14H10NO3
Molecular Weight (g/mol) 240.24
MDL Number MFCD00007715
InChI Key KORKIRUGUNPQML-UHFFFAOYSA-M
Synonym 2-2-aminobenzoyl benzoic acid, 2-aminobenzophenone-2'-carboxylic acid, 2-anthraniloylbenzoic acid, 2'-aminobenzophenone-2-carboxylic acid, benzoic acid,2-2-aminobenzoyl, 2-2-aminophenyl carbonyl benzoic acid, 2-aminobenzoyl benzoic acid, korkirugunpqml-uhfffaoysa, benzoic acid, 2-2-aminobenzoyl, 2-aminobenzophenone2'-carboxylic acid
PubChem CID 70847
IUPAC Name 2-(2-aminobenzoyl)benzoate
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1C([O-])=O

Specifications

Melting Point 199°C
Quantity 1 g
Formula Weight 241.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Aminobenzophenone-2-carboxylic Acid