accessibility menu, dialog, popup

UserName

NH2OOO

CAS RN 1147-43-9

IUPAC Name: 2-(2-aminobenzoyl)benzoate
Synonyms: 2-(2-aminobenzoyl)benzoate
Molecular Weight (g/mol): 240.24
Molecular Formula: C14H10NO3
InChi Key: KORKIRUGUNPQML-UHFFFAOYSA-M
SMILES: NC1=CC=CC=C1C(=O)C1=CC=CC=C1C([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results