missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenethiol 97.0+%, TCI America™

Supplier:  TCI America A0267500ML

 View more versions of this product

Catalog No. A0267500ML

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Aminobenzenethiol
Yellow
C6H7NS
500 mL
2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
NC1=CC=CC=C1S
125.19
125.19
Liquid
137-07-5
234°C
MFCD00007702
3267
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminobenzene-1-thiol
8713
≥97.0% (T)

Chemical Identifiers

137-07-5
125.19
VRVRGVPWCUEOGV-UHFFFAOYSA-N
8713
NC1=CC=CC=C1S
C6H7NS
MFCD00007702
2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
2-aminobenzene-1-thiol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2899

RTECSNumber : DC0600000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions