missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzenethiol 97.0+%, TCI America™

Catalog No. p-7116798
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A026725ML 25 mL
A0267100ML 100 mL
A0267500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A026725ML Supplier TCI America Supplier No. A026725ML
Only null left

Chemical Identifiers

CAS 137-07-5
Molecular Formula C6H7NS
Molecular Weight (g/mol) 125.19
MDL Number MFCD00007702
InChI Key VRVRGVPWCUEOGV-UHFFFAOYSA-N
Synonym 2-aminothiophenol, o-aminothiophenol, 2-mercaptoaniline, o-mercaptoaniline, o-aminobenzenethiol, 2-mercaptaniline, benzenethiol, 2-amino, 2-amino thiophenol, 1-amino-2-mercaptobenzene, 2-aminobenzene-1-thiol
PubChem CID 8713
IUPAC Name 2-aminobenzene-1-thiol
SMILES NC1=CC=CC=C1S

Specifications

Color Yellow
Boiling Point 234°C
Quantity 25 mL
UN Number 3267
Formula Weight 125.19
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Aminobenzenethiol
EINECSNumber (3)-2899
RTECSNumber DC0600000
TSCA Yes