missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoanthracene 98.0+%, TCI America™

Catalog No. A16871G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16871G Supplier TCI America Supplier No. A16871G

Chemical Identifiers

CAS 613-13-8
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00003582
InChI Key YCSBALJAGZKWFF-UHFFFAOYSA-N
Synonym 2-aminoanthracene, 2-anthramine, 2-anthracenamine, 2-anthrylamine, 2-anthracylamine, beta-aminoanthracene, 2-anthracenamide, ccris 22, .beta.-aminoanthracene, dsstox_cid_4458
PubChem CID 11937
ChEBI CHEBI:34260
IUPAC Name anthracen-2-amine
SMILES C1=CC=C2C=C3C=C(C=CC3=CC2=C1)N

Specifications

Quantity 1 g
Formula Weight 193.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Aminoanthracene
RTECSNumber CA9275000
TSCA No