accessibility menu, dialog, popup

UserName

H2N

CAS RN 613-13-8

IUPAC Name: anthracen-2-amine
Synonyms: 2-aminoanthracene 2-anthracylamine ccris 22
Molecular Weight (g/mol): 193.25
Molecular Formula: C14H11N
InChi Key: YCSBALJAGZKWFF-UHFFFAOYSA-N
SMILES: NC1=CC=C2C=C3C=CC=CC3=CC2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)