missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoanthracene, 94%

Catalog No. AAL1426903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1426903 1 g
AAL1426906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1426903 Supplier Thermo Scientific Chemicals Supplier No. L1426903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 613-13-8 | C14H11N | 193.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-13-8
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.249
MDL Number MFCD00003582
InChI Key YCSBALJAGZKWFF-UHFFFAOYSA-N
Synonym 2-aminoanthracene, 2-anthramine, 2-anthracenamine, 2-anthrylamine, 2-anthracylamine, beta-aminoanthracene, 2-anthracenamide, ccris 22, .beta.-aminoanthracene, dsstox_cid_4458
PubChem CID 11937
ChEBI CHEBI:34260
IUPAC Name anthracen-2-amine
SMILES C1=CC=C2C=C3C=C(C=CC3=CC2=C1)N

Specifications

Melting Point 235°C to 239°C
Quantity 1 g
Beilstein 2209414
Formula Weight 193.25
Percent Purity 94%
Chemical Name or Material 2-Aminoanthracene
Hazard Category H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 210-330-9
RTECSNumber CA9275000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only