missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-p-cresol Hydrochloride 98.0+%, TCI America™

Catalog No. A127625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A127625G 25 g
A1276500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A127625G Supplier TCI America Supplier No. A127625G

Chemical Identifiers

CAS 2977-71-1
Molecular Formula C7H10ClNO
Molecular Weight (g/mol) 159.613
MDL Number MFCD00191600
InChI Key MZMUKGFYWNOPAU-UHFFFAOYSA-N
Synonym 2-Amino-4-methylphenol Hydrochloride, 2-Hydroxy-5-methylaniline Hydrochloride
PubChem CID 3014155
IUPAC Name 2-amino-4-methylphenol;hydrochloride
SMILES CC1=CC(=C(C=C1)O)N.Cl

Specifications

Quantity 25 g
Formula Weight 159.61
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-p-cresol Hydrochloride