accessibility menu, dialog, popup

UserName

CAS RN 2977-71-1

HClCH3OHH2N

CAS RN 2977-71-1

IUPAC Name: hydrogen 2-amino-4-methylphenol chloride
Synonyms: hydrogen 2-amino-P-cresol chloride
Molecular Weight (g/mol): 159.61
Molecular Formula: C7H10ClNO
InChi Key: MZMUKGFYWNOPAU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=C(O)C(N)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results