missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-nitrobenzoic acid, 97%

Catalog No. p-7056894
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6609003 1 g
AAH6609006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6609003 Supplier Thermo Scientific Chemicals Supplier No. H6609003

CAS: 50573-74-5 | C7H6N2O4 | 182.135 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50573-74-5
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.135
MDL Number MFCD00034785
InChI Key GGKYLHNARFFORH-UHFFFAOYSA-N
Synonym 2-amino-6-nitrobenzoic acid, benzoic acid, 2-amino-6-nitro, 6-nitroanthranilic acid, 2-amino-6-nitro-benzoic acid, 2-amino-6-nitrobenzoicacid, 6-nitroanthanilic acid, acmc-209knw, 6-nitro-anthranilic acid, 6-nitro-2-aminobenzoesaure, ksc911o8d
PubChem CID 95795
SMILES C1=CC(=C(C(=C1)[N+](=O)[O-])C(=O)O)N

Specifications

Quantity 1 g
Beilstein 2807130
Formula Weight 182.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Amino-6-nitrobenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 610-543-0
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only