missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. A28475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A28475G 5 g
A28471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A28475G Supplier TCI America Supplier No. A28475G

Chemical Identifiers

CAS 53600-33-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD01941328
InChI Key DYZDIWNRWSNVPT-UHFFFAOYSA-N
Synonym 6-methoxyanthranilic acid, 6-amino-o-anisic acid, 2-amino-6-methoxy-benzoic acid, 2-amino-6-methoxybenzoicacid, benzoic acid, 2-amino-6-methoxy, 6-methoxy anthranilic acid, pubchem4706, 3-amino-2-carboxyanisole, 6-methoxy-anthranilic acid, 2-carboxy-3-methoxyaniline
PubChem CID 2735357
IUPAC Name 2-amino-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)N

Specifications

Melting Point 89°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxybenzoic Acid