accessibility menu, dialog, popup

UserName

H3COOHOH2N

CAS RN 53600-33-2

IUPAC Name: 2-amino-6-methoxybenzoic acid
Synonyms: 2-amino-6-methoxybenzoic acid
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: DYZDIWNRWSNVPT-UHFFFAOYSA-N
SMILES: COC1=CC=CC(N)=C1C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results