missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzoic acid, 97%

Catalog No. AAH3412803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3412803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3412803 Supplier Thermo Scientific Chemicals Supplier No. H3412803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 53600-33-2 | C8H9NO3 | 167.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 53600-33-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD01941328
InChI Key DYZDIWNRWSNVPT-UHFFFAOYSA-N
Synonym 6-methoxyanthranilic acid, 6-amino-o-anisic acid, 2-amino-6-methoxy-benzoic acid, 2-amino-6-methoxybenzoicacid, benzoic acid, 2-amino-6-methoxy, 6-methoxy anthranilic acid, pubchem4706, 3-amino-2-carboxyanisole, 6-methoxy-anthranilic acid, 2-carboxy-3-methoxyaniline
PubChem CID 2735357
IUPAC Name 2-amino-6-methoxybenzoic acid
SMILES COC1=CC=CC(=C1C(=O)O)N

Specifications

Melting Point ∽85°C (decomposition)
Quantity 1 g
Formula Weight 167.16
Percent Purity 97%
Chemical Name or Material 2-Amino-6-methoxybenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only