missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorobenzonitrile 98.0+%, TCI America™

Catalog No. A234425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A234425G 25 g
A23445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A234425G Supplier TCI America Supplier No. A234425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 77326-36-4
Molecular Formula C7H5FN2
Molecular Weight (g/mol) 136.13
MDL Number MFCD00015477
InChI Key IQUNZGOZUJITBJ-UHFFFAOYSA-N
Synonym 6-fluoroanthranilonitrile, 2-amino-6-fluorobenzo-nitrile, 2-fluoro-6-aminobenzonitrile, 6-fluoro-2-aminobenzonitrile, 2-amino-6-fluoro-benzonitrile, benzonitrile, 2-amino-6-fluoro, 2-amino-6-fluorobenzenecarbonitrile, pubchem4644, acmc-1bhvi, 2-cyano-3-fluoroaniline
PubChem CID 522659
IUPAC Name 2-amino-6-fluorobenzonitrile
SMILES NC1=CC=CC(F)=C1C#N

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 25 g
UN Number 3439
Formula Weight 136.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-fluorobenzonitrile
TSCA No
Recommended Storage Refrigerator