CAS RN 77326-36-4
IUPAC Name:
2-amino-6-fluorobenzonitrile
Synonyms:
2-amino-6-fluorobenzonitrile
Molecular Weight (g/mol):
136.13
Molecular Formula:
C7H5FN2
InChi Key:
IQUNZGOZUJITBJ-UHFFFAOYSA-N
SMILES:
NC1=CC=CC(F)=C1C#N
1
–
1
of
1
results
1
2-Amino-6-fluorobenzonitrile 98.0+%, TCI America™
CAS: 77326-36-4 Molecular Formula: C7H5FN2 Molecular Weight (g/mol): 136.13 MDL Number: MFCD00015477 InChI Key: IQUNZGOZUJITBJ-UHFFFAOYSA-N Synonym: 6-fluoroanthranilonitrile,2-amino-6-fluorobenzo-nitrile,2-fluoro-6-aminobenzonitrile,6-fluoro-2-aminobenzonitrile,2-amino-6-fluoro-benzonitrile,benzonitrile, 2-amino-6-fluoro,2-amino-6-fluorobenzenecarbonitrile,pubchem4644,acmc-1bhvi,2-cyano-3-fluoroaniline PubChem CID: 522659 IUPAC Name: 2-amino-6-fluorobenzonitrile SMILES: NC1=CC=CC(F)=C1C#N