missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzophenone 98.0+%, TCI America™

Catalog No. A113425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A113425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A113425G Supplier TCI America Supplier No. A113425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1775-95-7
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.23
MDL Number MFCD00007364
InChI Key PZPZDEIASIKHPY-UHFFFAOYSA-N
Synonym 2-amino-5-nitrobenzophenone, 2-amino-5-nitrophenyl phenyl methanone, methanone, 2-amino-5-nitrophenyl phenyl, 2-amino-5-nitrophenyl phenylmethanone, 2-amino-5-nitrophenyl-phenylmethanone, 2-benzoyl-4-nitroaniline, 5-nitro-2-aminobenzophenone, 2-amino-5-nitro-phenyl-phenyl-methanone, 2-amino-5-nitrophenyl phenyl ketone, pubchem3254
PubChem CID 15681
IUPAC Name 2-benzoyl-4-nitroaniline
SMILES NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O

Specifications

Melting Point 163°C
Color Yellow
Quantity 25 g
Formula Weight 242.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrobenzophenone
RTECSNumber PC4935000
TSCA No