accessibility menu, dialog, popup

UserName

NH2ONOO

CAS RN 1775-95-7

IUPAC Name: 2-benzoyl-4-nitroaniline
Synonyms: 2-amino-5-nitrobenzophenone
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: PZPZDEIASIKHPY-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)