missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzoic acid, 97%

Catalog No. p-7025786
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2145803 1 g
AAB2145806 5 g
AAB2145814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2145803 Supplier Thermo Scientific Chemicals Supplier No. B2145803

CAS: 446-08-2 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00055566
InChI Key FPQMGQZTBWIHDN-UHFFFAOYSA-N
Synonym 5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
PubChem CID 101412
ChEBI CHEBI:78042
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)N

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 2803664
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-fluorobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 207-159-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only