accessibility menu, dialog, popup

UserName

CAS RN 446-08-2

IUPAC Name: 2-amino-5-fluorobenzoic acid
Synonyms: 2-amino-5-fluorobenzoic acid
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: FPQMGQZTBWIHDN-UHFFFAOYSA-N
SMILES: NC1=CC=C(F)C=C1C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)