missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzoic acid, 97%

Catalog No. AC308940010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC308940010 1 g Glass bottle
AC308940050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC308940010 Supplier Thermo Scientific Chemicals Supplier No. 308940010

CAS: 446-08-2 | C7H6FNO2 | 155.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.13
MDL Number MFCD00055566
InChI Key FPQMGQZTBWIHDN-UHFFFAOYSA-N
Synonym 5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
PubChem CID 101412
ChEBI CHEBI:78042
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 181°C to 185°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula H2NC6H3(F)COOH
Quantity 1 g
Formula Weight 155.13
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Amino-5-fluorobenzoic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF
EINECSNumber 207-159-7

RUO – Research Use Only