Learn More
2-Amino-5-cyanopyridine, 98%
CAS: 4214-73-7 | C6H6N3 | 120.13 g/mol
$67.08 - $224.19
Chemical Identifiers
| CAS | 4214-73-7 |
|---|---|
| Molecular Formula | C6H6N3 |
| Molecular Weight (g/mol) | 120.13 |
| MDL Number | MFCD00128902 |
| InChI Key | KDVBYUUGYXUXNL-UHFFFAOYSA-O |
| Synonym | 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu |
| PubChem CID | 818260 |
| SMILES | NC1=CC=C(C=[NH+]1)C#N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1928206
|
Thermo Scientific Chemicals
L1928206 |
5 g |
Each for $67.08
|
|
|||||
|
AAL1928214
|
Thermo Scientific Chemicals
L1928214 |
25 g |
Each for $224.19
|
|
|||||
Description
2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Soluble in methanol. Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Chemical Identifiers
| 4214-73-7 | |
| 120.13 | |
| KDVBYUUGYXUXNL-UHFFFAOYSA-O | |
| 818260 |
| C6H6N3 | |
| MFCD00128902 | |
| 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu | |
| NC1=CC=C(C=[NH+]1)C#N |
Specifications
| 4214-73-7 | |
| 240°C to 250°C (15 mmHg) | |
| MFCD00128902 | |
| UN3439 | |
| 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu | |
| KDVBYUUGYXUXNL-UHFFFAOYSA-O | |
| 2-amino-5-cyanopyridin-1-ium | |
| 818260 | |
| 98% |
| 159°C to 164°C | |
| C6H6N3 | |
| 5 g | |
| 113876 | |
| Soluble in methanol. Insoluble in water. | |
| NC1=CC=C(C=[NH+]1)C#N | |
| 120.13 | |
| 119.13 | |
| 2-Amino-5-cyanopyridine |
Safety and Handling
GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only