accessibility menu, dialog, popup

UserName

NH2HNN

CAS RN 4214-73-7

IUPAC Name: 2-amino-5-cyanopyridin-1-ium
Synonyms: 2-amino-5-cyanopyridin-1-ium
Molecular Weight (g/mol): 120.13
Molecular Formula: C6H6N3
InChi Key: KDVBYUUGYXUXNL-UHFFFAOYSA-O
SMILES: NC1=CC=C(C=[NH+]1)C#N
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Boiling Point 
  • (2)
  • (2)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)
  • (4)