missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine, 98%

CAS: 4214-73-7 | C6H6N3 | 120.13 g/mol

Supplier:  Thermo Scientific Chemicals L1928214

 View more versions of this product

Catalog No. AAL1928214


Only null left
Explore available promotions

Description

Description

2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-Amino-5-cyanopyridine
159°C to 164°C
C6H6N3
25 g
113876
Soluble in methanol. Insoluble in water.
NC1=CC=C(C=[NH+]1)C#N
120.13
119.13
4214-73-7
240°C to 250°C (15 mmHg)
MFCD00128902
UN3439
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
KDVBYUUGYXUXNL-UHFFFAOYSA-O
6-aminopyridine-3-carbonitrile
818260
98%

Chemical Identifiers

4214-73-7
120.13
KDVBYUUGYXUXNL-UHFFFAOYSA-O
818260
C6H6N3
MFCD00128902
2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
NC1=CC=C(C=[NH+]1)C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H319
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H312-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only