missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-chlorophenol 97.0+%, TCI America™
Supplier: TCI America A19585G
Specifications
| 2-Amino-5-chlorophenol | |
| 155°C | |
| MFCD02093863 | |
| 4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol | |
| NC1=CC=C(Cl)C=C1O | |
| 143.57 | |
| CHEBI:75051 | |
| ≥97.0% (GC,T) |
| 28443-50-7 | |
| C6H6ClNO | |
| 5 g | |
| FZCQMIRJCGWWCL-UHFFFAOYSA-N | |
| 2-amino-5-chlorophenol | |
| 91591 | |
| 143.57 | |
| Crystalline Powder |
Chemical Identifiers
| 28443-50-7 | |
| 143.57 | |
| FZCQMIRJCGWWCL-UHFFFAOYSA-N | |
| 91591 | |
| 2-amino-5-chlorophenol |
| C6H6ClNO | |
| MFCD02093863 | |
| 4-chloro-2-hydroxyaniline, phenol, 2-amino-5-chloro, 2-amino-5-chloro-phenol, 5-chloro-2-aminophenol, pubchem1478, 5-chloro-o-aminophenol, 5-chloro-2-amino phenol, 2-hydroxy-4-chloroaniline, acmc-209h2j, 2-azanyl-5-chloranyl-phenol | |
| CHEBI:75051 | |
| NC1=CC=C(Cl)C=C1O |
Safety and Handling
EINECSNumber : (3)-0906
TSCA : Yes